Få mere at vide
1-Aminonaphthalene-d9, CDN
Artikelnummer. 30311439
Skift visning
Mængde:
0.05 g
0.1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30311439 0.05 g
30310024 0.1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30311439 Leverandør CDN Leverandørnr. CDND66850.05G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Aminonaphthalene-D9
CAS 78832-56-1
Molekylær formel C10 D9 N
Mængde 0.05 g
Synonym 1-Naphthalen-2,3,4,5,6,7,8-d7-amine-d2 (9 CI), 1-Napthylamine-d9, 1-Aminonaphthalene-d9, 1-Aminonaphthalene-D9, 1-Aminonaphthalene-N,N,2,3,4,5,6,7,8-d9
InChI formel InChI=1 S/C10H9N/c11-10-7-3-5-8-4-1-2-6-9(8)10/h1-7 H,11H2/i1D,2 D,3 D,4 D,5 D,6 D,7 D/hD2
SMIL [2 H]N([2 H])c1c([2 H])c([2 H])c([2 H])c2c([2 H])c([2 H])c([2 H])c([2 H])c12
IUPAC navn N,N,2,3,4,5,6,7,8-nonadeuterionaphthalen-1-amine
Molekylvægt (g/mol) 152.2406
Formel vægt 152.13 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity