Learn More
1-Aminoguanidinium Hydrogen Carbonate, Sigma-Aldrich™
Product Code. 15898530
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
Product Code. Quantity Unit Size
15898530 250 g 250g
1 options
MQ200 Graded - For Research and Non-regulated Industrial Applications
Product Code. 15898530 Supplier Sigma Aldrich Supplier No. 8014170250
 Restricted Item
Forventet forsendelse: 26-11-2026
MQ200 Graded - For Research and Non-regulated Industrial Applications
For synthesis
TRUSTED_SUSTAINABILITY
Specifications
CAS 2582-30-1
Melting Point 125°C (Decomposes)
Density 1.56 g/cm3 at 20°C
MDL Number MFCD00012949
Quantity 250 g
Synonym 1-Aminoguanidinium hydrogen carbonate, Aminoguanidine bicarbonate, 1-Aminoguanidinium hydrogen carbonate
Molecular Weight (g/mol) 136.11
Grade Research