missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-Aminocyclohexanecarboxylic acid hydrochloride, 98%

Catalog Number 11486946
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
39692-17-6
C7H14ClNO2
179.64
MFCD00044481
GTKXSYHXQSKWNP-UHFFFAOYSA-N
1-amino-1-cyclohexanecarboxylic acid hydrochloride, 1-aminocyclohexane-1-carboxylic acid hydrochloride, 1-aminocyclohexanecarboxylic acid hydrochloride, 1-amino-cyclohexanecarboxylic acid hcl, 1-aminocyclohexanecarboxylic acid, chloride, homocycloleucine hydrochloride, acmc-20aoyd, 1-aminocyclohexane-1-carboxylic acid hcl, 1-amino-cyclohexanecarboxylic acid hydrochloride, 1-aminocyclohexane-carboxylic acid hydrochloride
2724466
1-aminocyclohexane-1-carboxylic acid;hydrochloride
[H+].[Cl-].NC1(CCCCC1)C(O)=O
This item is not returnable. View return policy
39692-17-6
C7H14ClNO2
179.64
MFCD00044481
GTKXSYHXQSKWNP-UHFFFAOYSA-N
1-amino-1-cyclohexanecarboxylic acid hydrochloride, 1-aminocyclohexane-1-carboxylic acid hydrochloride, 1-aminocyclohexanecarboxylic acid hydrochloride, 1-amino-cyclohexanecarboxylic acid hcl, 1-aminocyclohexanecarboxylic acid, chloride, homocycloleucine hydrochloride, acmc-20aoyd, 1-aminocyclohexane-1-carboxylic acid hcl, 1-amino-cyclohexanecarboxylic acid hydrochloride, 1-aminocyclohexane-carboxylic acid hydrochloride
2724466
1-aminocyclohexane-1-carboxylic acid;hydrochloride
[H+].[Cl-].NC1(CCCCC1)C(O)=O
Melting Point >300°C (decomposition)
Quantity 25 g
Sensitivity Hygroscopic
Formula Weight 179.65
Percent Purity 98%
Chemical Name or Material 1-Aminocyclohexanecarboxylic acid hydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'