missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 1-Aminocyclohexanecarboxylic acid hydrochloride, 98%
Click to view available options
Quantity:
25 g
100 g
Specifications
Melting Point | >300°C (decomposition) |
Quantity | 100 g |
Sensitivity | Hygroscopic |
Formula Weight | 179.65 |
Percent Purity | 98% |
Chemical Name or Material | 1-Aminocyclohexanecarboxylic acid hydrochloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'