Få mere at vide
1-Amino-2-naphthol hydrochloride, 97%, Thermo Scientific™
Artikelnummer. 11435957
Skift visning
Mængde:
1 g
5 g
25 g
Pakningsstørrelse:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
11435957 1 g 1g
11445957 5 g 5g
11455957 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 11435957 Leverandør Thermo Scientific Alfa Aesar Leverandørnr. B24492.03
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1198-27-2
Molekylær formel C10H10ClNO
Molekylvægt (g/mol) 195.646
MDL nummer MFCD00012561
InChI nøgle DEKREBCFNLUULC-UHFFFAOYSA-N
Synonym 1-amino-2-naphthol hydrochloride, 1-aminonaphthalen-2-ol hydrochloride, 1-amino-2-naphthol hcl, unii-xm2lzu54ow, 2-naphthalenol, 1-amino-, hydrochloride 1:1, 2-naphthalenol, 1-amino-, hydrochloride, xm2lzu54ow, 1-aminonaphthalen-2-ol, chloride, timtec-bb sbb000347, acmc-209a4b
PubChem CID 518469
IUPAC navn 1-aminonaphthalen-2-ol;hydrochloride
SMIL C1=CC=C2C(=C1)C=CC(=C2N)O.Cl
Tekniske data
Kemisk navn eller materiale 1-Amino-2-naphthol hydrochloride
Smeltepunkt ∼276°C (decomposition)
Mængde 1 g
Beilstein 3699817
Følsomhed Light sensitive
Formel vægt 195.65
Procent renhed 97%