Få mere at vide
1-Amino-2,4-dibromo-anthraquinone, TRC
Artikelnummer. 30323301
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30323301 25 mg
30313897 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30323301 Leverandør Toronto Research Chemicals Leverandørnr. TRCA60446025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Amino-2,4-dibromo-anthraquinone
CAS 81-49-2
Molekylær formel C14H7Br2NO2
Mængde 25 mg
Synonym NSC 3529, 1-Amino-2,4-dibromo-9,10-anthracenedione, 1-Amino-2,4-dibromo-9,10-anthraquinone, 1-Amino-2,4-dibromoanthraquinone, 2,4-Dibromo-1-anthraquinonylamine
InChI formel InChI=1S/C14H7Br2NO2/c15-8-5-9(16)12(17)11-10(8)13(18)6-3-1-2-4-7(6)14(11)19/h1-5H,17H2
SMIL C1=CC2=C(C=C1)C(=O)C3=C(C(=CC(=C3C2=O)Br)Br)N
IUPAC navn 1-amino-2,4-dibromoanthracene-9,10-dione
Molekylvægt (g/mol) 381.02
Formel vægt 378.8844