Få mere at vide
1-Amino-1-cyclopentanecarboxylic Acid Methyl Ester, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 1-Amino-1-cyclopentanecarboxylic Acid Methyl Ester |
| CAS | 78388-61-1 |
| Molekylær formel | C7 H13 N O2 |
| Mængde | 100 mg |
| Synonym | Cyclopentanecarboxylic acid, 1-amino-, methyl ester, 1-Aminocyclopentanecarboxylic acid methyl ester, Cycloleucine methyl ester, Methyl 1-amino-1-cyclopentanecarboxylate, Methyl 1-aminocyclopentanecarboxylate, NSC 26981, 1-Amino-1-cyclopentanecarboxylic acid methyl ester |
| InChI formel | InChI=1S/C7H13NO2/c1-10-6(9)7(8)4-2-3-5-7/h2-5,8H2,1H3 |
| SMIL | COC(=O)C1(N)CCCC1 |
| IUPAC navn | methyl 1-aminocyclopentane-1-carboxylate |
| Molekylvægt (g/mol) | 143.18 |
| Formel vægt | 143.0946 |