Få mere at vide
1-(Allyloxy)-2-propanol, TRC
Artikelnummer. 30422564
Skift visning
Mængde:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30422564 500 mg
30416399 100 mg
30410777 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30422564 Leverandør Toronto Research Chemicals Leverandørnr. TRCP565255500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 21460-36-6
Molekylær formel C6H12O2
Molekylvægt (g/mol) 116.16
Synonym 1-(2-propen-1-yloxy)-2-Propanol, (9CI)1-(2-propenyloxy)-2-Propanol, (8CI)1-(allyloxy)-2-Propanol, 1-(2-Propen-1-yloxy)-2-propanol, 1-Allyloxypropane-2-ol, 2-(Allyloxy)-1-methylethanol, Allyl 2-hydroxypropyl ether
IUPAC navn 1-prop-2-enoxypropan-2-ol
SMIL CC(O)COCC=C
Tekniske data
Kemisk navn eller materiale 1-(Allyloxy)-2-propanol
Mængde 500 mg
InChI formel InChI=1S/C6H12O2/c1-3-4-8-5-6(2)7/h3,6-7H,1,4-5H2,2H3
Formel vægt 116.0837