missing translation for 'onlineSavingsMsg'
Learn More

1-Adamantyl methyl ketone, 99%, Thermo Scientific Chemicals

Catalog Number p-7037214
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
1660-04-4
C12H18O
178.28
MFCD00074739
DACIGVIOAFXPHW-UHFFFAOYSA-N
1-adamantyl methyl ketone, 1-acetyladamantane, 1-1-adamantyl ethanone, 1-adamantylmethylketone, 1-adamantan-1-yl ethan-1-one, 1-adamantan-1-yl ethanone, acetyladamantane, methyl 1-adamantyl ketone, 1-adamantan-1-yl-ethanone, adamantyl methyl ketone
123126
1-(1-adamantyl)ethanone
CC(=O)C12CC3CC(CC(C3)C1)C2
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
1660-04-4
C12H18O
178.28
MFCD00074739
DACIGVIOAFXPHW-UHFFFAOYSA-N
1-adamantyl methyl ketone, 1-acetyladamantane, 1-1-adamantyl ethanone, 1-adamantylmethylketone, 1-adamantan-1-yl ethan-1-one, 1-adamantan-1-yl ethanone, acetyladamantane, methyl 1-adamantyl ketone, 1-adamantan-1-yl-ethanone, adamantyl methyl ketone
123126
1-(1-adamantyl)ethanone
CC(=O)C12CC3CC(CC(C3)C1)C2

1-Adamantyl methyl ketone, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Adamantyl methyl ketone, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in methanol- very faint turbidity. Insoluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents. Stable under normal temperatures and pressures.
TRUSTED_SUSTAINABILITY
Melting Point 53°C to 55°C
Flash Point 108°C (226°F)
Quantity 1 g
Solubility Information Soluble in methanol- very faint turbidity. Insoluble in water.
Formula Weight 178.27
Percent Purity 99%
Chemical Name or Material 1-Adamantyl methyl ketone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'