missing translation for 'onlineSavingsMsg'
Learn More

1-Adamantyl isocyanate, 98%, Thermo Scientific Chemicals

Catalog Number p-3756541
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
4411-25-0
C11H15NO
177.25
MFCD00134405
VBHCPGFCIQDXGZ-UHFFFAOYSA-N
1-adamantyl isocyanate, 1-isocyanato-adamantane, 1-isocyantoadamantane, 1-adamantylisocyanate, adamantanisocyanate, adamantyl isocyanate, pubchem8848, 1-adamantyl-isocyanate, 1-adamantanyl isocyanate, adamantane, 1-isocyanato
2059995
1-isocyanatoadamantane
O=C=NC12CC3CC(CC(C3)C1)C2
This item is not returnable. View return policy
4411-25-0
C11H15NO
177.25
MFCD00134405
VBHCPGFCIQDXGZ-UHFFFAOYSA-N
1-adamantyl isocyanate, 1-isocyanato-adamantane, 1-isocyantoadamantane, 1-adamantylisocyanate, adamantanisocyanate, adamantyl isocyanate, pubchem8848, 1-adamantyl-isocyanate, 1-adamantanyl isocyanate, adamantane, 1-isocyanato
2059995
1-isocyanatoadamantane
O=C=NC12CC3CC(CC(C3)C1)C2

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 144.0°C to 146.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 177.25
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 1-Adamantyl isocyanate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'