Få mere at vide
1-Adamantan-d15-amine, TRC
Artikelnummer. 30317020
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30317020 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30317020 Leverandør Toronto Research Chemicals Leverandørnr. TRCA57582310MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Amantadine-d15
CAS 33830-10-3
Molekylær formel C10 2H15 H2 N
Mængde 10 mg
Synonym Amantadine D15, 1-Adamantan-d15-amine
InChI formel InChI=1S/C10H17N/c11-10-4-7-1-8(5-10)3-9(2-7)6-10/h7-9H,1-6,11H2/i1D2,2D2,3D2,4D2,5D2,6D2,7D,8D,9D
SMIL [2H]C1([2H])C2([2H])C([2H])([2H])C3([2H])C([2H])([2H])C1([2H])C([2H])([2H])C(N)(C3([2H])[2H])C2([2H])[2H]
IUPAC navn 2,2,3,4,4,5,6,6,7,8,8,9,9,10,10-pentadecadeuterioadamantan-1-amine
Molekylvægt (g/mol) 166.34
Formel vægt 166.2303