missing translation for 'onlineSavingsMsg'
Learn More

1-Acetylimidazole, 98%, Thermo Scientific Chemicals

Catalog Number 10683173
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
2466-76-4
C5H6N2O
110.12
MFCD00005287
VIHYIVKEECZGOU-UHFFFAOYSA-N
1-acetylimidazole, n-acetylimidazole, 1-1h-imidazol-1-yl ethanone, 1h-imidazole, 1-acetyl, acetylimidazole, 1-acetyl-1h-imidazole, imidazole, 1-acetyl, 3-acetylimidazole, unii-zmp8x1y11g, ccris 6534
17174
CHEBI:16984
1-imidazol-1-ylethanone
CC(=O)N1C=CN=C1
This item is not returnable. View return policy
2466-76-4
C5H6N2O
110.12
MFCD00005287
VIHYIVKEECZGOU-UHFFFAOYSA-N
1-acetylimidazole, n-acetylimidazole, 1-1h-imidazol-1-yl ethanone, 1h-imidazole, 1-acetyl, acetylimidazole, 1-acetyl-1h-imidazole, imidazole, 1-acetyl, 3-acetylimidazole, unii-zmp8x1y11g, ccris 6534
17174
CHEBI:16984
1-imidazol-1-ylethanone
CC(=O)N1C=CN=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 93°C to 96°C
Color Beige to White or Tan
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in ethanol,ether,chloroform,thf
Formula Weight 110.12
Percent Purity 98%
Physical Form Adhering Crystals or Crystalline Powder
Chemical Name or Material 1-Acetylimidazole, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'