missing translation for 'onlineSavingsMsg'
Learn More

1-Acetylimidazole-13C2, TRC

Product Code. 30325618
Change view
Click to view available options
Mængde:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30325618 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30325618 Supplier Toronto Research Chemicals Supplier No. TRCA179192100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 1-Acetylimidazole-13C2
CAS 2197068-07-6
Molekylær formel C313C2H6N2O
Mængde 100 mg
Synonym 1-(1H-Imidazol-1-yl)ethan-1-one-1,2-13C2
InChI formel InChI=1S/C5H6N2O/c1-5(8)7-3-2-6-4-7/h2-4H,1H3/i1+1,5+1
SMIL [13CH3][13C](=O)n1ccnc1
IUPAC navn 1-imidazol-1-yl(1,2-^{13}C_{2})ethanone
Molekylvægt (g/mol) 112.1
Formel vægt 112.0547
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.