missing translation for 'onlineSavingsMsg'
Få mere at vide

1-acetyl-N,N-diethyl-Lysergamide, TRC

Artikelnummer. 30004542
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30004542 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30004542 Leverandør Toronto Research Chemicals Leverandørnr. TRCL0030001MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale ALD-52
CAS 3270-02-8
Molekylær formel C23 H27 N O2
Mængde 1 mg
Synonym 1-Acetyllysergic Acid diethylamide, 1-Acetyl LSD
InChI formel InChI=1S/C23H27NO2/c1-4-24(5-2)23(26)16-10-9-15-11-17-13-20(14(3)25)18-7-6-8-19(22(17)18)21(15)12-16/h6-8,12-13,15-16,20H,4-5,9-11H2,1-3H3/t15?,16-,20?/m0/s1
SMIL CCN(CC)C(=O)[C@H]1CCC2CC3=CC(C(=O)C)c4cccc(C2=C1)c34
IUPAC navn (9S)-4-acetyl-N,N-diethyl-4,6,6a,7,8,9-hexahydroacephenanthrylene-9-carboxamide
Molekylvægt (g/mol) 349.46
Formel vægt 349.2042
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.