missing translation for 'onlineSavingsMsg'
Learn More

1-Acetyl-4-(methylamino)piperidine, Thermo Scientific Chemicals

Catalog Number p-7033811
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
139062-96-7
C8H16N2O
156.23
MFCD07367773
RSEPODZAQBVPOS-UHFFFAOYSA-N
1-acetyl-4-methylamino piperidine, 1-acetyl-n-methylpiperidin-4-amine, 1-4-methylamino piperidin-1-yl ethan-1-one, 1-4-methylamino piperidin-1-yl ethanone, 4-n-methylamino-1-acetyl-piperidine, 4-piperidinamine, 1-acetyl-n-methyl-9ci, 4-n-methylamino-1-acetyl, acmc-209vt3, 1-acetyl-4-methylaminopiperidine
19761265
CNC1CCN(CC1)C(C)=O
This item is not returnable. View return policy
139062-96-7
C8H16N2O
156.23
MFCD07367773
RSEPODZAQBVPOS-UHFFFAOYSA-N
1-acetyl-4-methylamino piperidine, 1-acetyl-n-methylpiperidin-4-amine, 1-4-methylamino piperidin-1-yl ethan-1-one, 1-4-methylamino piperidin-1-yl ethanone, 4-n-methylamino-1-acetyl-piperidine, 4-piperidinamine, 1-acetyl-n-methyl-9ci, 4-n-methylamino-1-acetyl, acmc-209vt3, 1-acetyl-4-methylaminopiperidine
19761265
CNC1CCN(CC1)C(C)=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
UN Number UN3259
IUPAC Name 1-[4-(methylamino)piperidin-1-yl]ethan-1-one
Formula Weight 156.23
Chemical Name or Material 1-Acetyl-4-(methylamino)piperidine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'