Få mere at vide
1'-Aceto-d3-naphthone, CDN
Artikelnummer. 30315488
Skift visning
Mængde:
0.5 g
1 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30315488 1 g -
30317972 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30315488 Leverandør CDN Leverandørnr. CDND81201G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Acetylnaphthalene-D3 (acetyl-D3)
CAS 95046-87-0
Molekylær formel C12 D3 H7 O
Mængde 1 g
Synonym Ethanone-2,2,2-d3, 1-(1-naphthalenyl)- (ACI), 1-(1-Naphthalenyl)ethanone-2,2,2-d3 (ACI), 1'-Acetonaphthone-2,2,2-d3 (7 CI), 1'-Aceto-d3-naphthone, 1-Acetylnaphthalene-d3, 1-Acetylnaphthalene-D3 (acetyl-D3), 1-Trideuterioacetylnaphthalene
InChI formel InChI=1 S/C12H10O/c1-9(13)11-8-4-6-10-5-2-3-7-12(10)11/h2-8 H,1H3/i1D3
SMIL [2 H]C([2 H])([2 H])C(=O)c1cccc2ccccc12
IUPAC navn 2,2,2-trideuterio-1-naphthalen-1-ylethanone
Molekylvægt (g/mol) 173.2257
Formel vægt 173.092 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity