Få mere at vide
1-Acetamido-4-cyano-3-methylisoquinoline, TRC
Artikelnummer. 30312889
Skift visning
Mængde:
50 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30312889 50 mg
30310545 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30312889 Leverandør Toronto Research Chemicals Leverandørnr. TRCA15052550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Acetamido-4-cyano-3-methylisoquinoline
CAS 179985-52-5
Molekylær formel C13 H11 N3 O
Mængde 50 mg
Synonym Acetamide, N-(4-cyano-3-methyl-1-isoquinolinyl)-, 1-Acetamido-4-cyano-3-methylisoquinoline, N-(4-Cyano-3-methyl-1-isoquinolinyl)acetamide
InChI formel InChI=1S/C13H11N3O/c1-8-12(7-14)10-5-3-4-6-11(10)13(15-8)16-9(2)17/h3-6H,1-2H3,(H,15,16,17)
SMIL CC(=O)Nc1nc(C)c(C#N)c2ccccc12
IUPAC navn N-(4-cyano-3-methylisoquinolin-1-yl)acetamide
Molekylvægt (g/mol) 225.25
Formel vægt 225.0902