Learn More
1-(9H-Fluoren-9-yl)-3-oxo-2,7,10-trioxa-4-azadodecan-12-oic acid, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 166108-71-0 |
| Molecular Formula | C21H23NO6 |
| MDL Number | MFCD01321015 |
| Quantity | 100 mg |
| SMILES | OC(=O)COCCOCCNC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2 |
| Molecular Weight (g/mol) | 385.41 |
| Percent Purity | 98% |
| Chemical Name or Material | 1-(9H-Fluoren-9-yl)-3-oxo-2,7,10-trioxa-4-azadodecan-12-oic acid |