Få mere at vide
1,9-Nonane-2,2,3,3,4,4,5,5,6,6,7,7,8,8-d14-diol, CDN
Artikelnummer. 30320599
Skift visning
Mængde:
0.1 g
0.25 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30320599 0.1 g 0.1g
30311015 0.25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30320599 Leverandør CDN Leverandørnr. CDND72230.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1,9-Nonanediol-2,2,3,3,4,4,5,5,6,6,7,7,8,8-D14
CAS 1219805-89-6
Molekylær formel C9 D14 H6 O2
Mængde 0.1 g
Synonym 1,9-Nonane-2,2,3,3,4,4,5,5,6,6,7,7,8,8-d14-diol, 1,9-Nonanediol-2,2,3,3,4,4,5,5,6,6,7,7,8,8-D14
InChI formel InChI=1 S/C9H20O2/c10-8-6-4-2-1-3-5-7-9-11/h10-11 H,1-9H2/i1D2,2D2,3D2,4D2,5D2,6D2,7D2
SMIL [2 H]C([2 H])(CO)C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])CO
IUPAC navn 2,2,3,3,4,4,5,5,6,6,7,7,8,8-tetradecadeuteriononane-1,9-diol
Molekylvægt (g/mol) 174.3402
Formel vægt 174.2342 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity