Få mere at vide
1,8-Octanedioic-d12 Acid, CDN
Artikelnummer. 30320294
Skift visning
Mængde:
0.1 g
0.25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30320294 0.25 g
30312245 0.1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30320294 Leverandør CDN Leverandørnr. CDND53380.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Octanedioic Acid-D12
CAS 169397-99-3
Molekylær formel C8 D12 H2 O4
Mængde 0.25 g
Synonym Octanedioic-2,2,3,3,4,4,5,5,6,6,7,7-d12 acid (9 CI), Suberic-2,2,3,3,4,4,5,5,6,6,7,7-d12 acid, 1,8-Octanedioic-2,2,3,3,4,4,5,5,6,6,7,7-d12 Acid, 1,6-Hexanedicarboxylic Acid-1,1,2,2,3,3,4,4,5,5,6,6-d12, Octanedioic acid-d12, Octanedioic Acid-D12, Suberic acid-d12
InChI formel InChI=1 S/C8H14O4/c9-7(10)5-3-1-2-4-6-8(11)12/h1-6H2,(H,9,10)(H,11,12)/i1D2,2D2,3D2,4D2,5D2,6D2
SMIL [2 H]C([2 H])(C(=O)O)C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C(=O)O
IUPAC navn 2,2,3,3,4,4,5,5,6,6,7,7-dodecadeuteriooctanedioic acid
Molekylvægt (g/mol) 186.2683
Formel vægt 186.1645 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity