missing translation for 'onlineSavingsMsg'
Learn More

1,8-Diiodoperfluorooctane, TRC

Product Code. 30377089
Change view
Click to view available options
Mængde:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30377089 1 g
30378944 250 mg
30385596 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30377089 Supplier Toronto Research Chemicals Supplier No. TRCD4942981G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 335-70-6
Molekylær formel C8F16I2
Molekylvægt (g/mol) 653.87
Synonym 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8-hexadecafluoro-1,8-diiodo-Octane, hexadecafluoro-1,8-diiodo-Octane, 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8-Hexadecafluoro-1,8-diiodooctane, 1,8-Diiodohexadecafluorooctane, alpha,ω-Diiodoperfluorooctane
SMIL FC(F)(I)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)I

Specifications

Kemisk navn eller materiale 1,8-Diiodoperfluorooctane
Mængde 1 g
InChI formel InChI=1S/C8F16I2/c9-1(10,3(13,14)5(17,18)7(21,22)25)2(11,12)4(15,16)6(19,20)8(23,24)26
Formel vægt 653.7834
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.