missing translation for 'onlineSavingsMsg'
Learn More

1,8-Dihydroxy-4,5-dinitroanthraquinone, 97%, Thermo Scientific™

Catalog Number p-7027952
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
81-55-0
C14H6N2O8
330.208
MFCD00019149
GJCHQJDEYFYWER-UHFFFAOYSA-N
1,8-dihydroxy-4,5-dinitroanthraquinone, 4,5-dinitrochrysazin, 1,8-dinitro-4,5-dihydroxyanthraquinone, 9,10-anthracenedione, 1,8-dihydroxy-4,5-dinitro, unii-heo6302d6d, ccris 4796, 4,5-dihydroxy-1,8-dinitroanthraquinone, anthraquinone, 1,8-dihydroxy-4,5-dinitro, 1,8-dihydroxy-4,5-dinitro-9,10-anthracenedione, 1,8-dihydroxy-4,5-dinitro-anthraquinone
3378440
1,8-dihydroxy-4,5-dinitroanthracene-9,10-dione
C1=CC(=C2C(=C1[N+](=O)[O-])C(=O)C3=C(C=CC(=C3C2=O)O)[N+](=O)[O-])O
This item is not returnable. View return policy
81-55-0
C14H6N2O8
330.208
MFCD00019149
GJCHQJDEYFYWER-UHFFFAOYSA-N
1,8-dihydroxy-4,5-dinitroanthraquinone, 4,5-dinitrochrysazin, 1,8-dinitro-4,5-dihydroxyanthraquinone, 9,10-anthracenedione, 1,8-dihydroxy-4,5-dinitro, unii-heo6302d6d, ccris 4796, 4,5-dihydroxy-1,8-dinitroanthraquinone, anthraquinone, 1,8-dihydroxy-4,5-dinitro, 1,8-dihydroxy-4,5-dinitro-9,10-anthracenedione, 1,8-dihydroxy-4,5-dinitro-anthraquinone
3378440
1,8-dihydroxy-4,5-dinitroanthracene-9,10-dione
C1=CC(=C2C(=C1[N+](=O)[O-])C(=O)C3=C(C=CC(=C3C2=O)O)[N+](=O)[O-])O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 225°C (decomposition)
Quantity 10 g
Formula Weight 330.21
Percent Purity 97%
Chemical Name or Material 1,8-Dihydroxy-4,5-dinitroanthraquinone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'