missing translation for 'onlineSavingsMsg'
Learn More

1,8-Diazido-3,6-dioxaoctane, TRC

Product Code. 30359794
Change view
Click to view available options
Mængde:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30359794 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30359794 Supplier Toronto Research Chemicals Supplier No. TRCD417200500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 1,8-Diazido-3,5-dioxaoctane
CAS 59559-06-7
Molekylær formel C6 H12 N6 O2
Mængde 500 mg
InChI formel InChI=1S/C6H12N6O2/c7-11-9-2-1-4-13-6-14-5-3-10-12-8/h1-6H2
SMIL [N-]=[N+]=NCCCOCOCCN=[N+]=[N-]
IUPAC navn 1-azido-3-(2-azidoethoxymethoxy)propane
Molekylvægt (g/mol) 200.2
Formel vægt 200.1022
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.