Få mere at vide
1,8-Di-p-toluidinoanthraquinone, TRC
Artikelnummer. 30133256
Skift visning
Mængde:
500 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30133256 5 g
30004409 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30133256 Leverandør Toronto Research Chemicals Leverandørnr. TRCD4946855G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 82-16-6
Molekylær formel C28H22N2O2
Molekylvægt (g/mol) 418.49
Synonym 1,8-Bis(p-methylphenylamino)anthraquinone, 1,8-di-p-toluidino-anthraquinone, 1,8-Di-p-toluidinoanthraquinone, 1,8-Bis[(4-methylphenyl)amino]-9,10-anthracenedione
IUPAC navn 1,8-Bis[(4-methylphenyl)amino]-9,10-anthracenedione
SMIL O=C1C=2C=CC=C(NC3=CC=C(C=C3)C)C2C(=O)C=4C(=CC=CC14)NC5=CC=C(C=C5)C
Tekniske data
Kemisk navn eller materiale 1,8-Di-p-toluidinoanthraquinone
Mængde 5 g
InChI formel InChI=1S/C28H22N2O2/c1-17-9-13-19(14-10-17)29-23-7-3-5-21-25(23)28(32)26-22(27(21)31)6-4-8-24(26)30-20-15-11-18(2)12-16-20/h3-16,29-30H,1-2H3
Formel vægt 418.17