Få mere at vide
1,7-Dimethyluric Acid-d3, TRC
Artikelnummer. 30373598
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30373598 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30373598 Leverandør Toronto Research Chemicals Leverandørnr. TRCD4796971MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1,7-Dimethyluric Acid-d3
CAS 1189713-08-3
Molekylær formel C7H5D3N4O3
Mængde 1 mg
InChI formel InChI=1S/C7H8N4O3/c1-10-3-4(8-6(10)13)9-7(14)11(2)5(3)12/h1-2H3,(H,8,13)(H,9,14)/i2D3
SMIL [2H]C([2H])([2H])N1C(=O)NC2=C(N(C)C(=O)N2)C1=O
Molekylvægt (g/mol) 199.18
Formel vægt 199.0785