Få mere at vide
1,7-Bis-(5-oxohexyl)-3-methylxanthine, Mikromol™
Artikelnummer. 30051069
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30051069 100 mg
30052124 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30051069 Leverandør Mikromol Leverandørnr. MM0875.03
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
1,7-Bis-(5-oxohexyl)-3-methylxanthine
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1,7-Bis-(5-oxohexyl)-3-methylxanthine
CAS 200556-62-3
Analyt- eller komponentnavne 1,7-Bis-(5-oxohexyl)-3-methylxanthine
Forsendelsestilstand Room Temperature
Molekylær formel C18 H26 N4 O4
Certificeringer/compliance ISO 9001
Synonym 1,7-Bis-(5-oxohexyl)-3-methylxanthine, Pentoxifylline Impurity H
InChI formel InChI=1S/C18H26N4O4/c1-13(23)8-4-6-10-21-12-19-16-15(21)17(25)22(18(26)20(16)3)11-7-5-9-14(2)24/h12H,4-11H2,1-3H3
SMIL CN1C(=O)N(CCCCC(=O)C)C(=O)c2c1ncn2CCCCC(=O)C
IUPAC navn 3-methyl-1,7-bis(5-oxohexyl)purine-2,6-dione
Molekylvægt (g/mol) 362.42
Mængde 100 mg
Formel vægt 362.1954 g/mol