missing translation for 'onlineSavingsMsg'
Learn More

1-(7-Amino-2-methyl-1-benzofuran-3-yl)ethanone, TRC

Product Code. 30325325
Change view
Click to view available options
Mængde:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30325325 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30325325 Supplier Toronto Research Chemicals Supplier No. TRCA133961500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 1-(7-Amino-2-methyl-1-benzofuran-3-yl)ethanone
CAS 174186-83-5
Molekylær formel C11H11NO2
Mængde 500 mg
InChI formel InChI=1S/C11H11NO2/c1-6(13)10-7(2)14-11-8(10)4-3-5-9(11)12/h3-5H,12H2,1-2H3
SMIL CC(=O)c1c(C)oc2c(N)cccc12
IUPAC navn 1-(7-amino-2-methyl-1-benzofuran-3-yl)ethanone
Molekylvægt (g/mol) 189.21
Formel vægt 189.079
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.