Få mere at vide
1-(6-Methyl-3-pyridinyl)-2-[4-(methylsulfonyl)phenyl]ethanone, Mikromol™
Artikelnummer. 17911136
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17911136 100 mg
17910946 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17911136 Leverandør Mikromol Leverandørnr. MM0965.05
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
1-(6-Methyl-3-pyridinyl)-2-[4-(methylsulfonyl)phenyl]ethanone
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-(6-Methyl-3-pyridinyl)-2-[4-(methylsulfonyl)phenyl]ethanone
CAS 221615-75-4
Analyt- eller komponentnavne 1-(6-Methyl-3-pyridinyl)-2-[4-(methylsulfonyl)phenyl]ethanone
Forsendelsestilstand Room Temperature
Molekylær formel C15 H15 N O3 S
Certificeringer/compliance ISO 9001
Synonym 1-(6-Methyl-3-pyridinyl)-2-[4-(methylsulfonyl)phenyl]-ethanone, 1-(6-Methylpyridin-3-yl)-2-[4-(methylsulfonyl)phenyl]ethanone
InChI formel InChI=1S/C15H15NO3S/c1-11-3-6-13(10-16-11)15(17)9-12-4-7-14(8-5-12)20(2,18)19/h3-8,10H,9H2,1-2H3
SMIL Cc1ccc(cn1)C(=O)Cc2ccc(cc2)S(=O)(=O)C
IUPAC navn 1-(6-methylpyridin-3-yl)-2-(4-methylsulfonylphenyl)ethanone
Molekylvægt (g/mol) 289.35
Mængde 100 mg
Formel vægt 289.0773 g/mol