Få mere at vide
1,6-Dioxacyclododecane-7,12-dione, TRC
Artikelnummer. 30003443
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30003443 10 mg
30004557 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30003443 Leverandør Toronto Research Chemicals Leverandørnr. TRCD48530510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 777-95-7
Molekylær formel C10H16O4
Molekylvægt (g/mol) 200.23
Synonym Adipic Acid Cyclic Tetramethylene Ester
SMIL O=C1OCCCCOC(CCCC1)=O
Tekniske data
Kemisk navn eller materiale 1,6-Dioxacyclododecane-7,12-dione
Renhedsgrad noter GC
Mængde 10 mg
InChI formel InChI=1S/C10H16O4/c11-9-5-1-2-6-10(12)14-8-4-3-7-13-9/h1-8H2
Formel vægt 200.1049
Procent renhed >95%