missing translation for 'onlineSavingsMsg'
Få mere at vide

1,6-Dihydroxynaphthalene, 99%

Artikelnummer. 11642919 Shop alle Thermo Scientific Chemicals produkter
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
25 g
100 g
Pakningsstørrelse:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
11642919 25 g 25g
11904681 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 11642919 Leverandør Thermo Scientific Acros Leverandørnr. 269910250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter

Denne vare kan ikke returneres. Se returpolicy

CAS: 575-44-0 | C10H8O2 | 160.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 575-44-0
Molekylær formel C10H8O2
Molekylvægt (g/mol) 160.17
InChI nøgle FZZQNEVOYIYFPF-UHFFFAOYSA-N
Synonym 1,6-dihydroxynaphthalene, 1,6-naphthalenediol, 6-hydroxy-1-naphthol, 1,6-dihydroxy naphthalene, 2,5-naphthalenediol, 2,5-dihydroxynaphthalene, naphthalene, 1,6-dihydroxy, ccris 7894, 1,6-dihydroxynaphtalene, 1.6-dihydroxynaphthalene
PubChem CID 68463
ChEBI CHEBI:42040
IUPAC navn naphthalene-1,6-diol
SMIL C1=CC2=C(C=CC(=C2)O)C(=C1)O

Tekniske data

Kemisk navn eller materiale 1, 6-Dihydroxynaphthalene
CAS min % 98.5
CAS Max % 100.0
Smeltepunkt 138.0°C to 140.0°C
Farve White to Brown
Infrarødt spektrum Authentic
Analyseprocentområde 98.5% min. (HPLC)
Emballage Glass bottle
Mængde 25 g
Formel vægt 160.17
Procent renhed 99%
Fysisk form Crystalline Powder or Powder
Vis mere Vis mindre
compliance-icons
Product Identifier
  • 1,6-Dihydroxynaphthalene
Signal Word
  • Advarsel
Hazard Category
  • Alvorlige øjenskader/øjenirritation Kategori 2
  • Hudætsning/hudirritation Kategori 2
  • Specifik målorgantoksicitet - enkelt eksponering Kategori 2
Hazard Statement
  • H319-Forårsager alvorlig øjenirritation.
  • H335-Kan forårsage irritation af luftvejene.
  • H315-Forårsager hudirritation.
Precautionary Statement
  • P261-Undgå indånding af pulver/røg/gas/tåge/damp/spray.
  • P280-Bær beskyttelseshandsker/beskyttelsestøj/øjenbeskyttelse/ ansigtsbeskyttelse.
  • P302+P352-VED KONTAKT MED HUDEN: Vask med rigeligt vand/.
  • P305+P351+P338-VED KONTAKT MED ØJNENE: Skyl forsigtigt med vand i flere minutter. Fjern eventuelle kontaktlinser, hvis dette kan gøres let. Fortsæt skylning.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.