Få mere at vide
1,6-Dihydro-5-hydroxy-1-methyl-2-[1-methyl-1-[[benzylcarbamoyl]amino]ethyl]-6-oxo-4-pyrimidinecarboxylic Acid Methyl Ester-d3, TRC
Artikelnummer. 30372773
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30372773 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30372773 Leverandør Toronto Research Chemicals Leverandørnr. TRCD4492421MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1,6-Dihydro-5-hydroxy-1-methyl-2-(1-methyl-1-((benzylcarbamoyl)amino)ethyl)-6-oxo-4-pyrimidinecarboxylic Acid Methyl Ester-d3
CAS 1189485-35-5
Molekylær formel C18H18D3N3O6
Mængde 1 mg
Synonym 4-Pyrimidinecarboxylic acid, 1,6-dihydro-5-hydroxy-1-methyl-2-[1-methyl-1-[[(phenylmethoxy)carbonyl]amino]ethyl]-6-oxo-, methyl ester (9CI, ACI), Methyl 1,6-dihydro-5-hydroxy-1-methyl-2-[1-methyl-1-[[(phenylmethoxy)carbonyl]amino]ethyl]-6-oxo-4-pyrimidinecarboxylate (ACI), 2-(1-Benzyloxycarbonylamino-1-methylethyl)-5-hydroxy-1-methyl-6-oxo-1,6-dihydropyrimidine-4-carboxylic acid Methyl Ester
InChI formel InChI=1S/C18H21N3O6/c1-18(2,20-17(25)27-10-11-8-6-5-7-9-11)16-19-12(15(24)26-4)13(22)14(23)21(16)3/h5-9,22H,10H2,1-4H3,(H,20,25)/i3D3
SMIL [2H]C([2H])([2H])N1C(=O)C(=C(N=C1C(C)(C)NC(=O)OCc2ccccc2)C(=O)OC)O
IUPAC navn methyl 5-hydroxy-6-oxo-2-[2-(phenylmethoxycarbonylamino)propan-2-yl]-1-(trideuteriomethyl)pyrimidine-4-carboxylate
Molekylvægt (g/mol) 378.39
Formel vægt 378.1619