Få mere at vide
1,6-Dichloronaphthalene, TRC
Artikelnummer. 30378923
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30378923 10 mg
30371575 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30378923 Leverandør Toronto Research Chemicals Leverandørnr. TRCD43666510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 2050-72-8
Molekylær formel C10H6Cl2
Molekylvægt (g/mol) 197.06
Synonym PCN 7
IUPAC navn 1,6-dichloronaphthalene
SMIL Clc1ccc2c(Cl)cccc2c1
Tekniske data
Kemisk navn eller materiale 1,6-Dichloronaphthalene
Mængde 10 mg
InChI formel InChI=1S/C10H6Cl2/c11-8-4-5-9-7(6-8)2-1-3-10(9)12/h1-6H
Formel vægt 195.9847