missing translation for 'onlineSavingsMsg'
Få mere at vide

1,6-Dibromohexane-d12, CDN

Artikelnummer. 30320903
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
0.1 g
0.25 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30320903 0.25 g -
30317494 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30320903 Leverandør CDN Leverandørnr. CDND37660.25G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale 1,6-Dibromohexane-D12
CAS 169397-96-0
Molekylær formel C6 D12 Br2
Mængde 0.25 g
Synonym Hexane-1,1,2,2,3,3,4,4,5,5,6,6-d12, 1,6-dibromo- (9 CI), 1,6-Dibromohexane-d12, 1,6-Dibromohexane-D12, 1,6-Dibromohexane-1,1,2,2,3,3,4,4,5,5,6,6-d12, Hexamethylene-d12 dibromide
InChI formel InChI=1 S/C6H12Br2/c7-5-3-1-2-4-6-8/h1-6H2/i1D2,2D2,3D2,4D2,5D2,6D2
SMIL [2 H]C([2 H])(Br)C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])Br
IUPAC navn 1,6-dibromo-1,1,2,2,3,3,4,4,5,5,6,6-dodecadeuteriohexane
Molekylvægt (g/mol) 256.0414
Formel vægt 254.0059 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity
Vis mere Vis mindre
Produkttitel
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.