Få mere at vide
1,6,7-Trimethyl-1H-imidazo[4,5-g]quinoxalin-2-amine-d3, TRC
Artikelnummer. 30453037
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30453037 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30453037 Leverandør Toronto Research Chemicals Leverandørnr. TRCT7960621MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1,6,7-Trimethyl-1H-imidazo[4,5-g]quinoxalin-2-amine-d3
CAS 1794759-37-7
Molekylær formel C12 D3 H10 N5
Mængde 1 mg
InChI formel InChI=1S/C12H13N5/c1-6-7(2)15-9-5-11-10(4-8(9)14-6)16-12(13)17(11)3/h4-5H,1-3H3,(H2,13,16)/i3D3
SMIL [2H]C([2H])([2H])n1c(N)nc2cc3nc(C)c(C)nc3cc12
IUPAC navn 6,7-dimethyl-3-(trideuteriomethyl)imidazo[4,5-g]quinoxalin-2-amine
Molekylvægt (g/mol) 230.284
Formel vægt 230.136