Få mere at vide
1,5-Pentane-d10-diol, TRC
Artikelnummer. 30421810
Skift visning
Mængde:
2.5 mg
5 mg
25 mg
Pakningsstørrelse:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30421810 5 mg -
30415323 2.5 mg 2.5mg
30411100 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30421810 Leverandør Toronto Research Chemicals Leverandørnr. TRCP2734065MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1219804-42-8
Molekylær formel C5 D10 H2 O2
Molekylvægt (g/mol) 114.209
Synonym 1,5-Pentane-1,1,2,2,3,3,4,4,5,5-d10-diol, 1,5-Pentane-d10-diol, 1,5-Pentanediol-d10, 1,5-Pentanediol-D10, 1,5-Dihydroxypentane-d10, 1,5-Pentamethylene-d10 glycol
IUPAC navn 1,1,2,2,3,3,4,4,5,5-decadeuteriopentane-1,5-diol
SMIL [2H]C([2H])(O)C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])O
Tekniske data
Kemisk navn eller materiale 1,5-Pentane-d10-diol
Mængde 5 mg
InChI formel InChI=1S/C5H12O2/c6-4-2-1-3-5-7/h6-7H,1-5H2/i1D2,2D2,3D2,4D2,5D2
Formel vægt 114.146