Få mere at vide
1-(5-O-Benzoyl-beta-D-ribofuranosyl)-1H-1,2,4-triazole-3-carboxamide (5'-O-Benzoylribavirin), Mikromol™
Artikelnummer. 30461844
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30461844 4 x 25 mg
30050910 25 mg
30050556 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30461844 Leverandør Mikromol Leverandørnr. MM0542.0500254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
1-(5-O-Benzoyl-beta-D-ribofuranosyl)-1H-1,2,4-triazole-3-carboxamide (5'-O-Benzoylribavirin), Mikromol
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 5'-O-Benzoylribavirin
CAS 58151-90-9
Analyt- eller komponentnavne 5'-O-Benzoylribavirin
Forsendelsestilstand Room Temperature
Molekylær formel C15 H16 N4 O6
Certificeringer/compliance ISO 17025
Synonym 1-(5-O-Benzoyl-beta-D-ribofuranosyl)-1H-1,2,4-triazole-3-carboxamide, 5'-O-Benzoylribavirin
InChI formel InChI=1S/C15H16N4O6/c16-12(22)13-17-7-19(18-13)14-11(21)10(20)9(25-14)6-24-15(23)8-4-2-1-3-5-8/h1-5,7,9-11,14,20-21H,6H2,(H2,16,22)/t9-,10-,11-,14-/m1/s1
Løsningstype Neat
SMIL NC(=O)c1ncn(n1)[C@@H]2O[C@H](COC(=O)c3ccccc3)[C@@H](O)[C@H]2O
IUPAC navn [(2R,3S,4R,5R)-5-(3-carbamoyl-1,2,4-triazol-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl benzoate
Molekylvægt (g/mol) 348.31
Mængde 4 x 25 mg
Formel vægt 348.107 g/mol