Få mere at vide
1-(5-Methylfuran-2-yl)propan-1-one, TRC
Artikelnummer. 30474540
Skift visning
Mængde:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30474540 50 mg
30482802 500 mg
30475283 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30474540 Leverandør Toronto Research Chemicals Leverandørnr. TRCM30580350MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 10599-69-6
Molekylær formel C8H10O2
Molekylvægt (g/mol) 138.16
Synonym 1-Propanone, 1-(5-methyl-2-furyl)- (6CI,7CI,8CI), 1-(5-Methyl-2-furanyl)-1-propanone1-(5-Methyl-2-furyl)propan-1-one, 2-Methyl-5-propionylfuran, 5-Methyl-2-propanoylfuran, 5-Methyl-2-propionylfuran
IUPAC navn 1-(5-methylfuran-2-yl)propan-1-one
SMIL CCC(=O)c1oc(C)cc1
Tekniske data
Kemisk navn eller materiale 1-(5-Methylfuran-2-yl)propan-1-one
Mængde 50 mg
InChI formel InChI=1S/C8H10O2/c1-3-7(9)8-5-4-6(2)10-8/h4-5H,3H2,1-2H3
Formel vægt 138.0681