missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,5-Dimethyl-1H-pyrazole-3-carboxylic acid, 95+%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
25 g
Specifications
Assay Percent Range | 95% |
Quantity | 1 g |
IUPAC Name | 1,5-dimethylpyrazole-3-carboxylic acid |
Formula Weight | 140.14 |
Percent Purity | 95% |
Chemical Name or Material | 1, 5-Dimethyl-1H-pyrazole-3-carboxylic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'