Learn More
1,5-Dichloro-2,4-dinitrobenzene, 97%, Thermo Scientific Chemicals
Catalog Number p-7044472
Click to view available options
Quantity:
5 g
25 g
3698-83-7
C6H2Cl2N2O4
236.992
MFCD00024186
ZPXDNSYFDIHPOJ-UHFFFAOYSA-N
1,3-dichloro-4,6-dinitrobenzene, benzene, 1,5-dichloro-2,4-dinitro, 2,4-dichloro-1,5-dinitrobenzene, 4,6-dichloro-1,3-dinitrobenzene, unii-6enc7c8zr0, 6enc7c8zr0, 1,5-dichloro-2,4-dinitro-benzene, 1,3,-dichloro-4,6-dinitrobenzene, 1,5-dichlor-2,4-dinitrobenzol, 2,5-dinitrobenzene
77286
1,5-dichloro-2,4-dinitrobenzene
C1=C(C(=CC(=C1[N+](=O)[O-])Cl)Cl)[N+](=O)[O-]
3698-83-7
C6H2Cl2N2O4
236.992
MFCD00024186
ZPXDNSYFDIHPOJ-UHFFFAOYSA-N
1,3-dichloro-4,6-dinitrobenzene, benzene, 1,5-dichloro-2,4-dinitro, 2,4-dichloro-1,5-dinitrobenzene, 4,6-dichloro-1,3-dinitrobenzene, unii-6enc7c8zr0, 6enc7c8zr0, 1,5-dichloro-2,4-dinitro-benzene, 1,3,-dichloro-4,6-dinitrobenzene, 1,5-dichlor-2,4-dinitrobenzol, 2,5-dinitrobenzene
77286
1,5-dichloro-2,4-dinitrobenzene
C1=C(C(=CC(=C1[N+](=O)[O-])Cl)Cl)[N+](=O)[O-]
1,5-Dichloro-2,4-dinitrobenzene is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,5-Dichloro-2,4-dinitrobenzene is used as pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 98°C to 103°C
Quantity 5 g
UN Number UN2811
Beilstein 1685438
Formula Weight 237
Percent Purity 97%
Chemical Name or Material 1,5-Dichloro-2,4-dinitrobenzene