Få mere at vide
1,5-Dibromopentane-1,1,5,5-d4, TRC
Artikelnummer. 30369440
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30369440 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30369440 Leverandør Toronto Research Chemicals Leverandørnr. TRCD42559125MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1,5-Dibromopentane-1,1,5,5-d4
CAS 1219803-90-3
Molekylær formel C5 D4 H6 Br2
Mængde 25 mg
Synonym 1,5-Dibromopentane-1,1,5,5-d4, 1,5-Dibromopentane-d4
InChI formel InChI=1S/C5H10Br2/c6-4-2-1-3-5-7/h1-5H2/i4D2,5D2
SMIL [2H]C([2H])(Br)CCCC([2H])([2H])Br
IUPAC navn 1,5-dibromo-1,1,5,5-tetradeuteriopentane
Molekylvægt (g/mol) 233.966
Formel vægt 231.94