Få mere at vide
1,5-Dibromopentane-1,1,5,5-d4, CDN
Artikelnummer. 30322318
Skift visning
Mængde:
0.1 g
0.25 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30322318 0.25 g -
30319010 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30322318 Leverandør CDN Leverandørnr. CDND60710.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1,5-Dibromopentane-1,1,5,5-d4
CAS 1219803-90-3
Molekylær formel C5 D4 H6 Br2
Mængde 0.25 g
Synonym 1,5-Dibromopentane-1,1,5,5-d4, 1,5-Dibromopentane-d4
InChI formel InChI=1 S/C5H10Br2/c6-4-2-1-3-5-7/h1-5H2/i4D2,5D2
SMIL [2 H]C([2 H])(Br)CCCC([2 H])([2 H])Br
IUPAC navn 1,5-dibromo-1,1,5,5-tetradeuteriopentane
Molekylvægt (g/mol) 233.966
Formel vægt 231.94 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity