Få mere at vide
1,5-Diaminonaphthalene-d6, TRC
Artikelnummer. 30349406
Skift visning
Mængde:
2.5 mg
Artikelnummer. Mængde
30349406 2.5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30349406 Leverandør Toronto Research Chemicals Leverandørnr. TRCD4164172.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1,5-Diaminonaphthalene-d6
CAS 1346598-98-8
Molekylær formel C10H4D6N2
Mængde 2.5 mg
Synonym 1,5-DAN-d6, 1,5-Diaminonaphthalene-d6, 1,5-Naphthylenediamine-d6, NSC 401110-d6
InChI formel InChI=1S/C10H10N2/c11-9-5-1-3-7-8(9)4-2-6-10(7)12/h1-6H,11-12H2/i1D,2D,3D,4D,5D,6D
SMIL NC1=C2C(C(N)=C([2H])C([2H])=C2[2H])=C([2H])C([2H])=C1[2H]
IUPAC navn naphthalene-d6-1,5-diamine
Molekylvægt (g/mol) 164.24
Formel vægt 164.12