Få mere at vide
1-[5-Chloro-2-(2,4-dichlorophenoxy)phenylethanone]-d3, TRC
Artikelnummer. 30367663
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30367663 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30367663 Leverandør Toronto Research Chemicals Leverandørnr. TRCC36524225MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-[5-Chloro-2-(2,4-dichlorophenoxy)phenylethanone]-d3
CAS 1189886-69-8
Molekylær formel C14 2H3 H6 Cl3 O2
Mængde 25 mg
InChI formel InChI=1S/C14H9Cl3O2/c1-8(18)11-6-9(15)2-4-13(11)19-14-5-3-10(16)7-12(14)17/h2-7H,1H3/i1D3
SMIL [2H]C([2H])([2H])C(=O)c1cc(Cl)ccc1Oc2ccc(Cl)cc2Cl
IUPAC navn 1-[5-chloro-2-(2,4-dichlorophenoxy)phenyl]-2,2,2-trideuterioethanone
Molekylvægt (g/mol) 318.6
Formel vægt 316.9856