Få mere at vide
1-[5-[(4,5-Diphenyl-1H-imidazol-2-yl)sulfinyl]pentyl]-3,5-dimethyl-1H-pyrazole, TRC
Artikelnummer. 30180224
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30180224 10 mg
30180136 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30180224 Leverandør Toronto Research Chemicals Leverandørnr. TRCD41530510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 136609-53-5
Molekylær formel C25 H28 N4 O S
Molekylvægt (g/mol) 432.581
Synonym RP 73163 Racemate
IUPAC navn 1-[5-[(4,5-diphenyl-1H-imidazol-2-yl)sulfinyl]pentyl]-3,5-dimethylpyrazole
SMIL Cc1cc(C)n(CCCCCS(=O)c2nc(c3ccccc3)c([nH]2)c4ccccc4)n1
Tekniske data
Kemisk navn eller materiale 1-[5-[(4,5-Diphenyl-1H-imidazol-2-yl)sulfinyl]pentyl]-3,5-dimethyl-1H-pyrazole
Mængde 10 mg
InChI formel InChI=1S/C25H28N4OS/c1-19-18-20(2)29(28-19)16-10-5-11-17-31(30)25-26-23(21-12-6-3-7-13-21)24(27-25)22-14-8-4-9-15-22/h3-4,6-9,12-15,18H,5,10-11,16-17H2,1-2H3,(H,26,27)
Formel vægt 432.198