Få mere at vide
1-[(4R,5R)-4,5-Dihydroxy-L-ornithine]-Echinocandin B Hydrochloride, TRC
Artikelnummer. 30383757
Skift visning
Mængde:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30383757 50 mg
30375627 500 mg
30386599 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30383757 Leverandør Toronto Research Chemicals Leverandørnr. TRCE32564550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1029890-89-8
Molekylær formel C34H51N7O15 . HCl
Molekylvægt (g/mol) 834.3
Synonym Anidulafungin Nucleus
SMIL Cl.C[C@@H](O)[C@@H]1NC(=O)C(NC(=O)[C@@H]2C[C@@H](O)CN2C(=O)C(NC(=O)[C@@H](N)C[C@@H](O)[C@@H](O)NC(=O)[C@@H]3[C@@H](O)[C@@H](C)CN3C1=O)[C@@H](C)O)[C@H](O)[C@@H](O)c4ccc(O)cc4
Tekniske data
Kemisk navn eller materiale 1-[(4R,5R)-4,5-Dihydroxy-L-ornithine]-Echinocandin B Hydrochloride
Mængde 50 mg
InChI formel InChI=1S/C34H51N7O15.ClH/c1-12-10-41-24(25(12)47)32(54)39-30(52)20(46)9-18(35)28(50)36-21(13(2)42)33(55)40-11-17(45)8-19(40)29(51)38-23(31(53)37-22(14(3)43)34(41)56)27(49)26(48)15-4-6-16(44)7-5-15;/h4-7,12-14,17-27,30,42-49,52H,8-11,35H2,1-3H3,(H,36,50)(H,37,53)(H,38,51)(H,39,54);1H/t12-,13+,14+,17+,18-,19-,20+,21-,22-,23-,24-,25-,26-,27-,30+;/m0./s1
Formel vægt 833.321