Learn More
1,4-Piperazinediethanesulfonic Acid Disodium Salt, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 76836-02-7 |
| Molecular Formula | C8H16N2Na2O6S2 |
| MDL Number | MFCD00064349 |
| Quantity | 100 g |
| SMILES | S(=O)(=O)([O-])CCN1CCN(CC1)CCS(=O)(=O)[O-].[Na+].[Na+] |
| Molecular Weight (g/mol) | 346.33 |
| Percent Purity | 98% |
| Chemical Name or Material | 1,4-Piperazinediethanesulfonic Acid Disodium Salt |