Få mere at vide
1-(4-O-Methoxymethyl-4-hydroxyphenyl)-4-(4-nitrophenyl)piperazine-d4, TRC
Artikelnummer. 30472570
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30472570 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30472570 Leverandør Toronto Research Chemicals Leverandørnr. TRCM264322100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-(4-O-Methoxymethyl-4-hydroxyphenyl)-4-(4-nitrophenyl)piperazine-d4
CAS 1246819-68-0
Molekylær formel C18 D4 H17 N3 O4
Mængde 100 mg
InChI formel InChI=1S/C18H21N3O4/c1-24-14-25-18-8-6-16(7-9-18)20-12-10-19(11-13-20)15-2-4-17(5-3-15)21(22)23/h2-9H,10-14H2,1H3/i2D,3D,4D,5D
SMIL [2H]c1c([2H])c(c([2H])c([2H])c1N2CCN(CC2)c3ccc(OCOC)cc3)[N+](=O)[O-]
IUPAC navn 1-[4-(methoxymethoxy)phenyl]-4-(2,3,5,6-tetradeuterio-4-nitrophenyl)piperazine
Molekylvægt (g/mol) 347.402
Formel vægt 347.178