missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Nitrophenyl)piperidine, 97%, Thermo Scientific™

Catalog Number 11484317
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
6574-15-8
C11H14N2O2
206.245
MFCD00023662
SGPLAXFUDTWHRS-UHFFFAOYSA-N
1-4-nitrophenyl piperidine, 1-p-nitrophenyl piperidine, n-4-nitrophenyl piperidine, 1-4-nitro-phenyl-piperidine, piperidine, 1-4-nitrophenyl, n-p-nitrophenyl piperidine, 4-nitrophenyl piperidine, pubchem9438, maybridge1_000682, acmc-20a12d
81049
1-(4-nitrophenyl)piperidine
C1CCN(CC1)C2=CC=C(C=C2)[N+](=O)[O-]
This item is not returnable. View return policy
6574-15-8
C11H14N2O2
206.245
MFCD00023662
SGPLAXFUDTWHRS-UHFFFAOYSA-N
1-4-nitrophenyl piperidine, 1-p-nitrophenyl piperidine, n-4-nitrophenyl piperidine, 1-4-nitro-phenyl-piperidine, piperidine, 1-4-nitrophenyl, n-p-nitrophenyl piperidine, 4-nitrophenyl piperidine, pubchem9438, maybridge1_000682, acmc-20a12d
81049
1-(4-nitrophenyl)piperidine
C1CCN(CC1)C2=CC=C(C=C2)[N+](=O)[O-]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 102°C to 105°C
Quantity 5 g
Formula Weight 206.24
Percent Purity 97%
Chemical Name or Material 1-(4-Nitrophenyl)piperidine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'