Learn More
1,4-Naphthoquinone, TRC
Product Code. 30479898
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
Product Code. Quantity Unit Size
30479898 25 g 25g
1 options
Product Code. 30479898 Supplier Toronto Research Chemicals Supplier No. TRCN36848825G
 Restricted Item
Forventet forsendelse: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1,4-Naphthoquinone
CAS 130-15-4
Molecular Formula C10 H6 O2
Quantity 25 g
Synonym 1,4-Naphthalenedione, 1,4-Naphthoquinone (8CI), 1,4-Dihydro-1,4-diketonaphthalene, 1,4-Naphthylquinone, NSC 113960, NSC 9583, Quino Power NQ, Quino Power NQI, p-Naphthoquinone, α-Naphthoquinone
InChI Formula InChI=1S/C10H6O2/c11-9-5-6-10(12)8-4-2-1-3-7(8)9/h1-6H
SMILES O=C1C=CC(=O)c2ccccc12
IUPAC Name naphthalene-1,4-dione
Molecular Weight (g/mol) 158.15
Formula Weight 158.0368