Få mere at vide
1-(4-Methylphenyl)-1-methylethylamine Hydrochloride, TRC
Artikelnummer. 30478566
Skift visning
Mængde:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30478566 500 mg
30473605 50 mg
30468121 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30478566 Leverandør Toronto Research Chemicals Leverandørnr. TRCM339295500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1654774-17-0
Molekylær formel C10H15N . HCl
Molekylvægt (g/mol) 185.69
IUPAC navn 2-(4-methylphenyl)propan-2-amine;hydrochloride
SMIL Cl.Cc1ccc(cc1)C(C)(C)N
Tekniske data
Kemisk navn eller materiale 1-(4-Methylphenyl)-1-methylethylamine Hydrochloride
Mængde 500 mg
InChI formel InChI=1S/C10H15N.ClH/c1-8-4-6-9(7-5-8)10(2,3)11;/h4-7H,11H2,1-3H3;1H
Formel vægt 185.0971